Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

163384

Sigma-Aldrich

4-(4-Methoxyphenyl)butyric acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4(CH2)3CO2H
CAS Number:
Molecular Weight:
194.23
Beilstein:
2416568
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

56-59 °C (lit.)

SMILES string

COc1ccc(CCCC(O)=O)cc1

InChI

1S/C11H14O3/c1-14-10-7-5-9(6-8-10)3-2-4-11(12)13/h5-8H,2-4H2,1H3,(H,12,13)

InChI key

LZHMNCJMXQKSBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674[1].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

163384-5G:
163384-500G:
163384-25G:
163384-VAR:
163384-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Demethylation of 4-methoxyphenylbutyric acid using molten pyridinium hydrochloride on multikilogram scale.
Schmid CR, et al.
Organic Process Research & Development, 8(4), 670-673 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service