Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

158070

Sigma-Aldrich

2-Oxopropyltriphenylphosphonium chloride

98%

Synonym(s):

Acetonyltriphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
354.81
Beilstein:
3922783
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

243-245 °C (lit.)

SMILES string

[Cl-].CC(=O)C[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H20OP.ClH/c1-18(22)17-23(19-11-5-2-6-12-19,20-13-7-3-8-14-20)21-15-9-4-10-16-21;/h2-16H,17H2,1H3;1H/q+1;/p-1

InChI key

XAMZZEBAJZJERT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used to investigate the antimycobacterial activities of Me alkenyl quinolones

Reactant for synthesis of:
  • Unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination
  • Fused oxygen and nitrogen heterocycles via regioselective cyclocondensation

Reactant for Wittig olefinations

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

158070-BULK:
158070-VAR:
158070-100G:
158070-25G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service