Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

15540

Sigma-Aldrich

Boc-Lys(Z)-OH (dicyclohexylammonium) salt

≥99.0% (NT)

Synonym(s):

Nα-Boc-Nε-Z-L-lysine (dicyclohexylammonium) salt, Nε-Z-Nα-Boc-L-lysine (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28N2O6 · C12H23N
CAS Number:
Molecular Weight:
561.75
Beilstein:
4121117
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (NT)

form

solid

optical activity

[α]20/D −5.2±0.5°, c = 1% in acetic acid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

ign. residue

≤0.05%

mp

114-116 °C

application(s)

peptide synthesis

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCc3ccccc3)C(O)=O

InChI

1S/C19H28N2O6.C12H23N/c1-19(2,3)27-18(25)21-15(16(22)23)11-7-8-12-20-17(24)26-13-14-9-5-4-6-10-14;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h4-6,9-10,15H,7-8,11-13H2,1-3H3,(H,20,24)(H,21,25)(H,22,23);11-13H,1-10H2/t15-;/m0./s1

InChI key

BQERJWRZLXZNIO-RSAXXLAASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

15540-5G:
15540-VAR:
15540-BULK:
15540-25G:
15540-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service