Skip to Content
Merck
All Photos(1)

Documents

Safety Information

154245

Sigma-Aldrich

4-Bromo-2-chloroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)NH2
CAS Number:
Molecular Weight:
206.47
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

70-72 °C (lit.)

SMILES string

Nc1ccc(Br)cc1Cl

InChI

1S/C6H5BrClN/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

INMZDDDQLHKGPF-UHFFFAOYSA-N

Application

4-Bromo-2-chloroaniline was used in the peparation of 3,4-dichloro-α-trichloromethylbenzyl alcohol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

154245-VAR:
154245-10G:
154245-BULK:
154245-50G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE REACTION OF CHLORAL WITH HALOBENZENES AND WITH 2, 4-DICHLOROPHENOL: SYNTHESIS OF HALOGENATED ETHYLBENZENES AND OF α-METHOXYPHENYLACETIC ACIDS.
Reeve W, et al.
Canadian Journal of Chemistry, 44(5), 575-582 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service