Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

150428

Sigma-Aldrich

3-Chloro-2-methylpropionitrile

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(CH3)CN
CAS Number:
Molecular Weight:
103.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

granular

refractive index

n20/D 1.434 (lit.)

bp

60 °C/15 mmHg (lit.)

density

1.078 g/mL at 25 °C (lit.)

SMILES string

CC(CCl)C#N

InChI

1S/C4H6ClN/c1-4(2-5)3-6/h4H,2H2,1H3

InChI key

XXZVBSPSFVERCB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

150428-100G:4548173106106
150428-BULK:
150428-VAR:
150428-25G:4548173106113


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service