Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

148474

Sigma-Aldrich

1,2,3-Tribromopropane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(Br)CH2Br
CAS Number:
Molecular Weight:
280.78
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.584 (lit.)

bp

220 °C (lit.)

mp

16-17 °C (lit.)

solubility

H2O: insoluble
alcohol: soluble
chloroform: soluble
diethyl ether: soluble

density

2.398 g/mL at 25 °C (lit.)

SMILES string

BrCC(Br)CBr

InChI

1S/C3H5Br3/c4-1-3(6)2-5/h3H,1-2H2

InChI key

FHCLGDLYRUPKAM-UHFFFAOYSA-N

General description

1,2,3-Tribromopropane was mutagenic in strains TA1535 and TA100 of Salmonella typhimurium.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>248.0 °F - closed cup - does not flash

Flash Point(C)

> 120 °C - closed cup - does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

148474-VAR:
148474-5G:
148474-100G:
148474-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Ratpan et al.
Environmental and molecular mutagenesis, 12(2), 253-259 (1988-01-01)
1,2,3-Tribromopropane, 1,2,3-trichloropropane, and 1,2-dibromo-3-chloropropane are mutagenic in strains TA1535 and TA100 of Salmonella typhimurium, but only in the presence of rat liver S9 mix. This requirement for metabolic activation was unexpected for an alkyl halide and thus suggested the metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service