Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

147850

Sigma-Aldrich

2-Cyclopenten-1-one ethylene ketal

95%

Synonym(s):

1,4-Dioxaspiro[4.4]non-6-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

59-62 °C/16 mmHg (lit.)

density

1.067 g/mL at 25 °C (lit.)

functional group

ether
ketal

storage temp.

2-8°C

SMILES string

C1COC2(CCC=C2)O1

InChI

1S/C7H10O2/c1-2-4-7(3-1)8-5-6-9-7/h1,3H,2,4-6H2

InChI key

ATFWVBMLTLPOHI-UHFFFAOYSA-N

Application

2-Cyclopenten-1-one ethylene ketal was used in the synthesis of 2,3,3,4,7,7-hexahydro-5-methyl-ind-5-en-1-one ethylene ketal.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

147850-VAR:
147850-25G:
147850-BULK:
147850-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of methyl epijasmonate and cis-3-(2-oxopropyl)-2-(pent-2-Z-enyl)-cyclopentan-1-one.
Hailes HC, et al.
Tetrahedron, 57(52), 10329-10333 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service