Skip to Content
Merck
All Photos(1)

Documents

144355

Sigma-Aldrich

4-Aminodibenzenesulfonamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O4S2
CAS Number:
Molecular Weight:
312.36
MDL number:

Assay

99%

SMILES string

Nc1ccc(cc1)S(=O)(=O)NS(=O)(=O)c2ccccc2

InChI

1S/C12H12N2O4S2/c13-10-6-8-12(9-7-10)20(17,18)14-19(15,16)11-4-2-1-3-5-11/h1-9,14H,13H2

InChI key

TXWROWPXUQZXFZ-UHFFFAOYSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service