Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

142174

Sigma-Aldrich

6-Ethoxy-2-mercaptobenzothiazole

99%

Synonym(s):

6-Ethoxy-2-benzothiazolethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NOS2
CAS Number:
Molecular Weight:
211.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

198-200 °C (lit.)

SMILES string

CCOc1ccc2nc(S)sc2c1

InChI

1S/C9H9NOS2/c1-2-11-6-3-4-7-8(5-6)13-9(12)10-7/h3-5H,2H2,1H3,(H,10,12)

InChI key

HOASVNMVYBSLSU-UHFFFAOYSA-N

General description

6-Ethoxy-2-mercaptobenzothiazole is a potential matrix for matrix-assisted laser desorption mass spectrometry for accurate mass determination of biomolecules[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

142174-50G:
142174-BULK:
142174-5G:
142174-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercaptobenzothiazoles: A new class of matrices for laser desorption ionization mass spectrometry.
Xu N, et al.
Journal of the American Society For Mass Spectrometry, 8(2), 116-124 (1997)
Fen Pei et al.
Scientific reports, 7(1), 17803-17803 (2017-12-21)
Quantitative Systems Pharmacology (QSP) is a drug discovery approach that integrates computational and experimental methods in an iterative way to gain a comprehensive, unbiased understanding of disease processes to inform effective therapeutic strategies. We report the implementation of QSP to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service