Skip to Content
Merck
All Photos(1)

Documents

Safety Information

11252

Sigma-Aldrich

cis-2-Aminocycloheptanecarboxylic acid hydrochloride

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO2 · HCl
CAS Number:
Molecular Weight:
193.67
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

product name

cis-2-Aminocycloheptanecarboxylic acid hydrochloride, ≥98.0% (TLC)

Assay

≥98.0% (TLC)

form

powder or crystals

color

colorless to white

application(s)

peptide synthesis

SMILES string

Cl.N[C@@H]1CCCCC[C@@H]1C(O)=O

InChI

1S/C8H15NO2.ClH/c9-7-5-3-1-2-4-6(7)8(10)11;/h6-7H,1-5,9H2,(H,10,11);1H/t6-,7+;/m0./s1

InChI key

KCRZBDJVYOBHIP-UOERWJHTSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

11252-500MG-F:
11252-BULK-F:
11252-VAR-F:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service