ADVH996C0FCA
AChemBlock
2-Ethyl-2-methyl-decanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
214.35
Iupac Name
2-ethyl-2-methyldecanoic acid
Inchi Code
1S/C13H26O2/c1-4-6-7-8-9-10-11-13(3,5-2)12(14)15/h4-11H2,1-3H3,(H,14,15)
InChI key
JFWZRVKAMYDLHQ-UHFFFAOYSA-N
Purity
0.97
Country of Origin
CN
Physical Form
colorless oil
Product Link
https://www.achemblock.com/x188559-2-ethyl-2-methyl-decanoic-acid.html
Synonym
2-ethyl-2-methyldecanoic acid
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/x188559-2-ethyl-2-methyl-decanoic-acid.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.