Skip to Content
Merck
All Photos(1)

Key Documents

PHL83533

Quercetin 3-gentiobioside

phyproof® Reference Substance

Synonym(s):

Quercetin 3-O-β-gentiobioside, 3,3′,4′,5,7-Pentahydroxyflavone 3-gentiobioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O17
CAS Number:
Molecular Weight:
626.52
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OC[C@H]2O[C@H]([C@@H]([C@H]([C@@H]2O)O)O)Oc3[c](c4c([o]c3c5cc(c(cc5)O)O)cc(cc4O)O)=O

InChI key

FDRQPMVGJOQVTL-DEFKTLOSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: primula

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service