Skip to Content
Merck
All Photos(1)

Documents

FAP79867

Nut allergen in biscuit

reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

Fera Science Ltd

application(s)

food and beverages

format

matrix material

storage temp.

−20°C

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Allergens
Hazelnut and Peanut. Value for whole hazelnut and whole peanut protein spike and reference values for different ELISA kits.

Matrix Group: Processed Food

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service