Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501126

Sigma-Aldrich

Anti-TNKS (N-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PARP-5a, Anti-PARP5A, Anti-PARPL, Anti-TIN1, tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNKS(8658)

General description

The gene TNKS (tankyrase) is mapped to human chromosome 8p23.1. It belongs to the PARP (poly (ADP-ribose) polymerase) family of proteins and is responsible for addition of poly (ADP-ribose) (PAR) to substrate proteins. The TNKS protein contains ankyrin repeats and a sterile α-motif (SAM).

Immunogen

Peptide with sequence RECOMBINANT, from the N Terminal Region of the protein sequence according to NP_003738.2.

Biochem/physiol Actions

TNKS (tankyrase) is a regulator of Wnt-signaling. It negatively regulates Axin1, thereby suppressing β-catenin degradation and causing hyperactivation of Wnt/β-catenin signaling. TNKS controls cell death and telomere length. It might also be associated with osteoarthritis. In murine lung cancer model, silencing of this gene reduces lung cancer cell growth. Application of TNKS inhibitor suppresses growth of hepatocellular carcinoma cells. Absence of TNKS activity helps in replication of human cytomegalovirus (HCMV).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic variants in eleven telomere-associated genes and the risk of incident cardio/cerebrovascular disease: The Women's Genome Health Study.
Zee RY, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 412, 199-202 (2011)
A genome-wide association study of copy-number variation identifies putative loci associated with osteoarthritis in Koreans.
Moon S, et al.
BMC Musculoskelet. Disord., 16, 76-76 (2015)
Tankyrase 1 polymorphism associated with an increased risk in developing non-small cell lung cancer in a Chinese population: a proof-of-principle study.
Wang Y, et al.
International Journal of Clinical and Experimental Pathology, 8, 10500-10511 (2015)
Tankyrase inhibitors attenuate WNT/?-catenin signaling and inhibit growth of hepatocellular carcinoma cells.
Ma L, et al.
Oncotarget, 6, 25390-25401 (2015)
Sujayita Roy et al.
Viruses, 8(1) (2016-01-06)
Human cytomegalovirus (HCMV) was reported to downregulate the Wnt/β-catenin pathway. Induction of Axin1, the negative regulator of the Wnt pathway, has been reported as an important mechanism for inhibition of β-catenin. Since Tankyrase (TNKS) negatively regulates Axin1, we investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service