Skip to Content
Merck
All Photos(2)

Key Documents

SAB1404611

Sigma-Aldrich

Monoclonal Anti-TNKS antibody produced in mouse

clone 1A3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.99 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNKS(8658)

General description

Mouse monoclonal antibody raised against a partial recombinant TNKS.
The gene TNKS (tankyrase) is mapped to human chromosome 8p23.1. It belongs to the PARP (poly (ADP-ribose) polymerase) family of proteins and is responsible for addition of poly (ADP-ribose) (PAR) to substrate proteins. The TNKS protein contains ankyrin repeats and a sterile α-motif (SAM).

Immunogen

TNKS (NP_003738, 1017 a.a. ~ 1124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TERKEGEVAGLDMNISQFLKSLGLEHLRDIFETEQITLDVLADMGHEELKEIGINAYGHRHKLIKGVERLLGGQQGTNPYLTFHCVNQGTILLDLAPEDKEYQSVEEE

Biochem/physiol Actions

TNKS (tankyrase) is a regulator of Wnt-signaling. It negatively regulates Axin1, thereby suppressing β-catenin degradation and causing hyperactivation of Wnt/β-catenin signaling. TNKS controls cell death and telomere length. It might also be associated with osteoarthritis. In murine lung cancer model, silencing of this gene reduces lung cancer cell growth. Application of TNKS inhibitor suppresses growth of hepatocellular carcinoma cells. Absence of TNKS activity helps in replication of human cytomegalovirus (HCMV).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sujayita Roy et al.
Viruses, 8(1) (2016-01-06)
Human cytomegalovirus (HCMV) was reported to downregulate the Wnt/β-catenin pathway. Induction of Axin1, the negative regulator of the Wnt pathway, has been reported as an important mechanism for inhibition of β-catenin. Since Tankyrase (TNKS) negatively regulates Axin1, we investigated the
Genetic variants in eleven telomere-associated genes and the risk of incident cardio/cerebrovascular disease: The Women's Genome Health Study.
Zee RY, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 412, 199-202 (2011)
Tankyrase inhibitors attenuate WNT/?-catenin signaling and inhibit growth of hepatocellular carcinoma cells.
Ma L, et al.
Oncotarget, 6, 25390-25401 (2015)
A genome-wide association study of copy-number variation identifies putative loci associated with osteoarthritis in Koreans.
Moon S, et al.
BMC Musculoskelet. Disord., 16, 76-76 (2015)
Tankyrase 1 polymorphism associated with an increased risk in developing non-small cell lung cancer in a Chinese population: a proof-of-principle study.
Wang Y, et al.
International Journal of Clinical and Experimental Pathology, 8, 10500-10511 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service