HUMANHL34-0002235
Frozen Human Normal CD34+ Hematopoietic Stem Cells
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Synonym(s):
HLA typed, Hematopoietic Stem Cells, Immune Cells, Mobilized CD34+ Cells
About This Item
product name
Frozen Human Normal CD34+ Hematopoietic Stem Cells, 20 Million Cells/per vial- HLA Typed
biological source
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Quality Level
form
frozen liquid
packaging
vial of 20 Million cells
manufacturer/tradename
BioIVT
technique(s)
cell culture | mammalian: suitable
color
cloudy
storage temp.
-140 to -196°C
General description
We are pleased to offer these products from BioIVT.
Hematopoietic stem cells (HSCs) are progenitor cells that, through hematopoeisis, differentiate into blood cells. CD34+ HSCs are found in the peripheral blood, bone marrow, and cord blood. CD34+ HSCs are separated from mononuclear cells via immunomagnetic separation.
Used for a variety of applications, HSCs are frequently utilized in cell therapy tissue engineering, transplant research, and other immunology applications.
Packaging
Other Notes
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service