Key Documents
EP5490042000
Rotor for Eppendorf® Concentrator Plus
F-50-8-18, non aerosol tight, to hold, 8 x 10/18 mL round-bottome tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 8 x 10/18 mL round-bottome tubes
to hold 8 x 18 x 105 - 128 mm
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490042000 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
General description
Fixed angle rotor F-50-8-18 can hold a maximum of 8 x 10/18 mL round-bottomed tubes with diameter of 105 - 128 mm. It is not aerosol tight and is to be used with Eppendorf Concentrator Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG