AXYPCRAS200
Aluminum sealing film for thermal and general sealing
size 35 μm, pkg of 5x100sheets/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Recommended Products
material
aluminum
sterility
non-sterile
feature
piercable
manufacturer/tradename
Axygen Inc PCR-AS-200
Corning
packaging
pkg of 5x100sheets/cs
size
35 μm
Related Categories
General description
- Aluminum sealing films are ideal for light-sensitive samples.
- Uniform adhesive enables efficient well sealing.
- Versatile sealing film can be used in various applications including PCR, Real-Time PCR to ELISA and cell culture.
- Compatible with a wide range of reagents and temperatures.
- Axygen® aluminum sealing films are ideal for light-sensitive samples.
- Uniform adhesive enables efficient well sealing.
- Versatile sealing film can be used in various applications including PCR, Real-Time PCR to ELISA and cell culture.
- Compatible with a wide range of reagents and temperatures.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)