VHPPWRSPJP
Milliflex oasis® VHP resistant pump power supply
Milliflex Oasis® VHP pump power supply for JP
Synonym(s):
Milliflex Oasis® VHP pump power supply - JP, To connect up to 2 filtration pumps
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122409
packaging
pkg of 1 ea
manufacturer/tradename
Milliflex®
technique(s)
Rapid Sterility Testing: suitable
application(s)
bioburden testing
pharmaceutical
sterility testing
compatibility
for use with Milliflex Oasis® VHP vacuum pump
Looking for similar products? Visit Product Comparison Guide
General description
Milliflex Oasis® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. Milliflex Oasis® VHP resistant pump is a new generation vacuum pump for sterility and bioburden analysis in pharmaceutical environments. A power supply of European standard with 100 - 240 V, 50/60 enables efficient functioning of the filtration pump. One set of Milliflex Oasis® power supply can be used to power up to two pumps.
Application
Milliflex Oasis® VHP resistant pump finds its application in bioburden testing and sterility testing for Pharmaceutical Products Analysis.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service