Skip to Content
Merck
All Photos(1)

Key Documents

MXRPWRSPUK

Millipore

Milliflex® Rapid System 2.0 power supply

Milliflex Oasis pumppower supply for United Kingdom, for use with the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station

Synonym(s):

Milliflex® Rapid System 2.0 power supply

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
JA.13

packaging

pkg of 1 ea

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

application(s)

bioburden testing
pharmaceutical
sterility testing

compatibility

for use with the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station

Looking for similar products? Visit Product Comparison Guide

General description

The Milliflex® Rapid System 2.0 is an automated solution for the rapid detection, response and resolution of microbial contamination in filterable samples throughout the manufacturing process. Milliflex® Rapid System 2.0 power supply is set with of European standard with 100 - 240 V, 50/60 enables efficient functioning of the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station.

Application

Milliflex® Rapid System 2.0 finds its application in sterility testing and bioburden testing for Pharmaceutical Products Analysis.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service