Skip to Content
Merck
All Photos(1)

Documents

MAB2511Z

Sigma-Aldrich

Anti-VCAM-1 Antibody, clone P8B1, Azide Free

clone P8B1, Chemicon®, from mouse

Synonym(s):

CD106, MAB2511

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

P8B1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

MAB2511Z recognizes the human VCAM-1 molecule. The antibody inhibits VLA-4 (integrin alpha4beta1) binding and recognizes a different epitope than that which is recognized by MAB2144 (P3C4 clone).

Application

Anti-VCAM-1 Antibody, clone P8B1, Azide Free is an antibody against VCAM-1.
Function blocking

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Liquid in 0.2 M phosphate buffer to 0.25 M NaCl, pH 7.6, containing no preservatives.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service