Skip to Content
Merck
All Photos(2)

Key Documents

KMEFHAW10S

Millipore

EZ-Fit® Manifold Kit

Contains 4 boxes of EZ-Fit™ 100 mL Funnels with single packaging, EZ-Fit® 3-head Manifold with filtration heads

Synonym(s):

EZ-Fit® Filtration system for bioburden testing, EZ-Fit® Manifold Starting Kit, Filtration Kit, Filtration Unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

Agency

ISO 7004

sterility

electron beam irradiated

packaging

pkg of 4 units (multi packaging)

manufacturer/tradename

EZ-Fit®

parameter

45 °C temperature (for samples)

volume

100 mL

pore size

0.45 μm pore size

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

shipped in

ambient

General description

EZ-Fit® Manifold Starting Kit is designed in a way to support laboratory filtration test methods. The kit contains EZ-Fit® Manifold and 4 boxes of EZ-Fit® 100ml Funnels making it the perfect kit to start sample testing through filtration.
The EZ-Fit® Filtration device is designed to fit onto EZ-Fit® Manifold, which features specific EZ-Fit® Filtration Heads.
The state-of-the-art design is optimized to secure laboratory workflow, avoid cross contamination and save testing time thereby producing reliable microbiological results.
After the filtration process is completed, the membrane can be easily transferred onto an agar plate for microbial growth.

Application

EZ-Fit® Manifold Starting Kit can be used to carry out bioburden testing of liquid samples like water, process samples or final products.

Packaging

EFHAW10MS x4 – 192 units, Multipack packaging
EZFITBASE3 x1
EZFITMVHE3 x1

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service