1.46562
Tryptic Soy Broth Ready-to-use Bottles
bottle capacity 500 mL, bottle filling volume 500 mL, closure type, blue crimp cap with septum, box of 6 bottles
Synonym(s):
CASO, Casein Soybean Digest Broth, SCDB, TSB, Tryptic Soy Broth, Tryptone Soy Broth
About This Item
Recommended Products
Agency
EP 2.6.1
JP 4.06
USP 71
71
Quality Level
sterility
sterile; autoclaved
form
liquid
feature
closure type blue crimp cap with septum
ready-to-use
packaging
box of 6 bottles
bottle capacity
500 mL
bottle filling volume
500 mL
pH
7.3±0.2
application(s)
cosmetics
food and beverages
pharmaceutical
sterility testing
storage temp.
2-25°C
suitability
nonselective for
Looking for similar products? Visit Product Comparison Guide
General description
The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, that promotes the growth of the microbes present in the test sample.
Application
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service