Skip to Content
Merck
All Photos(3)

Key Documents

1.06645

SAFC

Sodium sulfate

anhydrous, fine powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, JPE, USP

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9
Pricing and availability is not currently available.

Agency

BP
JPE
Ph. Eur.
USP

Quality Level

product line

EMPROVE® ESSENTIAL

form

fine powder

particle size

(see specification)

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium sulfate anhydrous is the anhydrous, sodium salt form of sulfuric acid, and can be used as buffer and pH modifier or mineral salt.
Sodium sulfate anhydrous fine powder Emprove® Essential is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service