Z279994
Norprene® A-60-G laboratory tubing
I.D. × O.D. 3/8 in. × 9/16 in.
Synonym(s):
Saint Gobain AFL00028, Tygon A-60-G
All Photos(1)
About This Item
UNSPSC Code:
41105108
NACRES:
NB.51
Recommended Products
packaging
pkg of 50 feet
manufacturer/tradename
Tygon
I.D. × O.D.
3/8 in. × 9/16 in.
wall size
3/32 in.
Related Categories
General description
This chemical pump tubing is a thermoplastic elastomer tubing with chemical compatibilities approaching fluoroelastomers and silicone tubing. Black, rubber-like tubing is heat sealable. Max. temp.: 275°F (135°C).
- Chemical resistance to oxidizing agents, acids, and alkalies
- Resistance to flex-fatigue, will not weaken or crack
Legal Information
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)