description
no. of flexible bonds 32
no. of rigid bonds 100
feature
no. of atoms 64
General description
Large, lightweight molecular models for classroom demonstration of organic and inorganic structures. For building models to scale of 20 cm/Å and 8 cm/Å. Bond lengths range from 5.5 to 27 cm. Atomic centers are 5 cm in diameter, finished in standard atom colors, and have a reinforced vinyl receptor at each bond position for insertion of connecting rods (bonds). All model kits include storage box and instructions. Additional atoms and bonds are available separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)