Key Documents
SYNP170005
Synthware round bottom pressure vessel with PTFE bushing
Synthware™ round bottom pressure vessel with PTFE bushing
350 mL, O.D. × L 80 mm × 154 mm, #15 bushing
Synonym(s):
Pressure Vessel, Synthware P170005, pressure tube
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P170005
O.D. × L
80 mm × 154 mm
capacity
350 mL
fitting
#15 bushing
General description
Heavy wall round bottom vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Active Filters
    • Review 1
    • Votes 0
    3 out of 5 stars.
    Synthware™ round bottom pressure vessel with PTFE
    It is a good equipment of glass regarding its quality however I could not find and data related to pressure rate so I do not how far I can go with temperature. Solvent is water.
    Helpful?
    1. Response from MilliporeSigma:
      Thank you for your review. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.