SLW1526/16D
Pyrex® Reagent bottles, unground narrow neck
capacity 5000 mL
Synonym(s):
Pyrex, SciLabware 1526/16D
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
Pyrex bottle
colorless bottle
narrow-mouth bottle
Joint
Joints ST/NS neck (44)
feature
without cap
packaging
pack of 1 ea
manufacturer/tradename
SciLabware 1526/16D
bottle diam. × H
187 mm × 315 mm
capacity
5000 mL
neck I.D.
44 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with ISO 4796
- Borosilicate glass with unground neck
- For use with rubber stoppers (not supplied)
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service