GF67785856
Silicon carbide
powder, 75 max. part. size (micron), weight 50 g, purity 98.7%
Synonym(s):
SI676010
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101521
NACRES:
NA.23
Recommended Products
Assay
98.7%
form
powder
manufacturer/tradename
Goodfellow 677-858-56
weight
50 g
max. part. size
75 (micron)
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service