GF46411017
Nimonic® alloy 90, Ni53Cr20Co18Ti2.5Al1.5Fe1.5
rod, 200mm x 22mm diameter
Synonym(s):
NJ107950
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
rod
manufacturer/tradename
Goodfellow 464-110-17
L × diam.
200 mm × 22 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Nimonic is a registered trademark of Special Metals Corporation
Nimonic is a registered trademark of Special Metals Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service