CLS3838BC
Corning® 1536 well plate, with generic barcode
size 1536 wells, flat bottom clear polystyrene wells, sterile, lid
Synonym(s):
1536 multiwell plate, 1536 well barcode plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, 1536 well plate with generic barcode
About This Item
flat bottom clear polystyrene wells
polystyrene plate
Recommended Products
material
black plate
flat bottom clear polystyrene wells
polystyrene plate
sterility
sterile
feature
lid
skirt
packaging
pack of 10
pkg of 10 ea
case of 50 ea
case of 50
manufacturer/tradename
Corning 3838BC
maximum volume
12.5 μL
size
1536 wells
working volume
8 μL
color
clear
suitability
suitable for (cell cuture)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
- Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
- Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
- Bar Code labeling specifications
- 2.75" x 0.25" label size
- Code 128
- Code 3 of 9
- 10 mil narrow bar element (x-dimension = 0.010")
- Resistant to temperature ranges of -80ºC to +52ºC
- 2.8 mm distance between well bottoms to plate bottom
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
Microplate Features
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Superior flatness -minimizes variability within a plate and between plates
- Flat bottom wells are compatible with automated dispensers
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²)
- Working volume of up to 10 μL
Bar Code Details
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
- Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
- Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
- Bar Code labeling specifications
- 2.75" x 0.25" label size
- Code 128
- Code 3 of 9
- 10 mil narrow bar element (x-dimension = 0.010")
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service