Skip to Content
Merck
All Photos(2)

Documents

329274

Sigma-Aldrich

2-Methoxy-5-nitrophenol

98%

Synonym(s):

5-Nitroguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)OH
CAS Number:
Molecular Weight:
169.13
Beilstein:
2047074
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

110-112 °C/1 mmHg (lit.)

mp

103-107 °C (lit.)

SMILES string

COc1ccc(cc1O)[N+]([O-])=O

InChI

1S/C7H7NO4/c1-12-7-3-2-5(8(10)11)4-6(7)9/h2-4,9H,1H3

InChI key

KXKCTSZYNCDFFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxy-5-nitrophenol is the substitution photoproduct formed on irradiation of 2-chloro-4-nitroanisole at 25°C in aqueous NaOH.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene G Wubbels et al.
The Journal of organic chemistry, 78(10), 4834-4839 (2013-05-02)
Irradiation (λ > 330 nm) of 2-chloro-4-nitroanisole (1) at 25 °C in aqueous NaOH forms three substitution photoproducts: 2-methoxy-5-nitrophenol (2), 2-chloro-4-nitrophenol (3), and 3-chloro-4-methoxyphenol (4), in chemical yields of 69.2%, 14.3%, and 16.5%. The activation energies for the elementary steps
Nenad Manevski et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 126-139 (2014-10-24)
Although skin is the largest organ of the human body, cutaneous drug metabolism is often overlooked, and existing experimental models are insufficiently validated. This proof-of-concept study investigated phase II biotransformation of 11 test substrates in fresh full-thickness human skin explants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service