Skip to Content
Merck
All Photos(1)

Key Documents

321710

Sigma-Aldrich

Carbon dioxide

10% in helium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352119

concentration

10% in helium

Application

For calibrating analytical instruments.

Packaging

Packaged in lightweight no-return cylinders

Recommended products

Control valve Z166995 or regulator Z167002 is recommended.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service