Skip to Content
Merck
All Photos(1)

Key Documents

SML2689

Sigma-Aldrich

EN6

≥98% (HPLC)

Synonym(s):

N-(3-Acrylamido-4-fluorophenyl)-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide, N-[4-Fluoro-3-[(1-oxo-2-propen-1-yl)amino]phenyl]-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14F2N4O2
CAS Number:
Molecular Weight:
368.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=C(NC(C=C)=O)C=C(NC(C2=CN(C3=C(F)C=CC=C3)N=C2)=O)C=C1

InChI key

SUSXQEYPNDORDQ-UHFFFAOYSA-N

Biochem/physiol Actions

Autophagy activator that upregulates lysosomal v-ATPase activity and induces its decoupling from Rag GTPases by covalently targeting v-ATPase subunit ATP6V1A Cys277.
EN6 is an autophagy activator that simultaneously upregulates lysosomal vacuolar H(+)-ATPase (v-ATPase) activity and induces its decoupling from the Rag GTPases by covalently targeting v-ATPase subunit ATP6V1A Cys277. EN6 blocks mTORC1 lysosomal recruitment & activation (1-4 hr 25 μM EN6 pretreatment prior to 10-min amino acids (AA) stimulation of AA-starived HEK293) and clears cellular TDP-43 aggregates (25 μM; U2OS with inducible TDP-43). When administered in mice in vivo (50 mg/kg ip.), EN6 inhibits mTORC1 signaling and activates autophagy in both skeletal muscle and heart tissue. Unlike bafilomycin A1 (BafA1), EN6 activates, but not inhibits v-ATPase catalytic activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clive Yik-Sham Chung et al.
Nature chemical biology, 15(8), 776-785 (2019-07-10)
Autophagy is a lysosomal degradation pathway that eliminates aggregated proteins and damaged organelles to maintain cellular homeostasis. A major route for activating autophagy involves inhibition of the mTORC1 kinase, but current mTORC1-targeting compounds do not allow complete and selective mTORC1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service