Skip to Content
Merck
All Photos(1)

Documents

COR-L47

92031915, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR-L47, 92031915

biological source

human lung

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

High levels of L-dopa decarboxylase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian small cell lung carcinoma

Cell Line Description

Derived from the bone marrow aspirate of a 67-year-old Caucasian male suffering from small cell lung carcinoma.

Application

Studies on lung cancer

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 11
D16S539: 11,12
D5S818: 10,13
D7S820: 8,10
THO1: 6,8
TPOX: 8
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells are difficult to count as they form large clumps. Cells grow as floating aggregates and become confluent within 2 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service