Skip to Content
Merck
All Photos(1)

Documents

MDA-MB-361

92020423, human breast (brain metastasis), Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

MDA-MB-361, 92020423

biological source

human breast (brain metastasis)

growth mode

Adherent

karyotype

Modal no. 56

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian breast adenocarcinoma

Cell Line Description

This cell line originates from a 40 year old caucasian female suffering from a breast adenocarcinoma but these cells were isolated from a metastatic site in the brain.

Application

MDA-MB-361 has been used to study the effects of low-intensity cytotoxic ultrasound irradiation for induction of cell ablation.

Culture Medium

L15 + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e. seeding at 3-6x10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 37°C. CO2 is not required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service