Key Documents
ADF356309S-EP
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 x 9 - 15 mL round-bottom tube and blood collection tube, small rotor bore, small rotor bore, pack of 2 ea
About This Item
UNSPSC Code:
41103911
feature
holds 1 x 9 - 15 mL round-bottom tube and blood collection tube, small rotor bore
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 022654538 (NA)
parameter
7,745 × g max. RCF
technique(s)
centrifugation: suitable
diam.
17 mm
compatibility
for use with Rotor F-35-6-30 (small rotor bore)
General description
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)