PHR2994
Aminolevulinic Acid Related Compound B
pharmaceutical secondary standard, certified reference material
Synonym(s):
1,3-Dioxo-2-isoindolinelevulinic acid methyl ester, Methyl 5-(1,3-dioxoisoindolin-2-yl)-4-oxopentanoate
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1025737
API family
aminolevulinic acid
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
N1(C(=O)c2c(cccc2)C1=O)CC(=O)CCC(=O)OC
InChI
1S/C14H13NO5/c1-20-12(17)7-6-9(16)8-15-13(18)10-4-2-3-5-11(10)14(15)19/h2-5H,6-8H2,1H3
InChI key
KFILPVALWMTVMP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service