Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 60 g
manufacturer/tradename
NIST®
format
matrix material
General description
This Standard Reference Material (SRM) is a fine powder of borate ore and is intended for use in the evaluation of chemical and instrumental methods of analyses. For more information, please refer to the SDS and COA.
SRM 1835_cert
SRM 1835_SDS
SRM 1835_cert
SRM 1835_SDS
Other Notes
Certified for the analytes listed below.
Silicon dioxide (SiO2), Aluminum oxide (Al2O3), Boron trioxide (B2O3), Barium oxide (BaO), Calcium oxide (CaO), Fluorine (F), Iron (III) Oxide (Fe2O3), Potassium oxide (K2O), Magnesium oxide (MgO), Manganese(II) oxide (MnO), Sodium oxide (Na2O), Sulfur trioxide (SO3), Strontium oxide (SrO), Titanium dioxide (TiO2)
Reference/Informational value is provided for the following analytes. Silicon dioxide (SIO2), Aluminum oxide (Al2O3), Boron trioxide (B2O3), Barium oxide (BaO), Calcium oxide (CaO), Fluorine (F), Iron (III) Oxide (Fe2O3), Potassium oxide (K2O), Magnesium oxide (MgO), Manganese(II) oxide (MnO), Sodium oxide (Na2O), Sulfur trioxide (SO3), Strontium oxide (SrO), Titanium dioxide (TiO2)
See certificate for values and more details at nist.gov/SRM.
Silicon dioxide (SiO2), Aluminum oxide (Al2O3), Boron trioxide (B2O3), Barium oxide (BaO), Calcium oxide (CaO), Fluorine (F), Iron (III) Oxide (Fe2O3), Potassium oxide (K2O), Magnesium oxide (MgO), Manganese(II) oxide (MnO), Sodium oxide (Na2O), Sulfur trioxide (SO3), Strontium oxide (SrO), Titanium dioxide (TiO2)
Reference/Informational value is provided for the following analytes. Silicon dioxide (SIO2), Aluminum oxide (Al2O3), Boron trioxide (B2O3), Barium oxide (BaO), Calcium oxide (CaO), Fluorine (F), Iron (III) Oxide (Fe2O3), Potassium oxide (K2O), Magnesium oxide (MgO), Manganese(II) oxide (MnO), Sodium oxide (Na2O), Sulfur trioxide (SO3), Strontium oxide (SrO), Titanium dioxide (TiO2)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service