Skip to Content
Merck
All Photos(1)

Key Documents

24585

Hamilton® GASTIGHT® syringe, 1700 series

1701N, volume 10 μL, needle size 26s ga (side-port), needle L 51 mm (2 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
Pricing and availability is not currently available.

material

PTFE plunger tip
cone tip, side port stainless steel needle (point style 5)

needle size

26s ga (side-port)

description

Gastight®

feature

needle type N (cemented)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80039

needle L

51 mm (2 in.)

volume

10 μL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service