Skip to Content
Merck
All Photos(1)

Key Documents

05-4950

Sigma-Aldrich

Citric acid monohydrate

≥99.5%, suitable for amino acid analysis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2 · H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
4018641
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

<0.01 hPa ( 25 °C)

Assay

≥99.5%

form

solid

availability

available only in Japan

pH

1.85 (25 °C, 50 g/L)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

suitability

suitable for amino acid analysis

SMILES string

OC(CC(O)(C(O)=O)CC(O)=O)=O.O

InChI

1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2

InChI key

YASYEJJMZJALEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

345.0 °F - closed cup

Flash Point(C)

173.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Ganss et al.
PloS one, 10(6), e0123889-e0123889 (2015-06-16)
For preventing erosive wear in dentine, coating with adhesives has been suggested as an alternative to fluoridation. However, clinical studies have revealed limited efficacy. As there is first evidence that Sn(2+) increases bond strength of the adhesive Clearfil SE (Kuraray)
T Ulens et al.
Journal of animal science, 93(2), 627-636 (2014-12-31)
This study evaluated the effect of feed form and grinding intensity of the pig diet and the interaction between both on the particulate matter (PM) concentrations inside a pig nursery and the growth performances of weanling pigs. Four diets were
Toine F H Bovee et al.
Mutation research, 778, 1-10 (2015-05-30)
It has previously been demonstrated by others that acetone extracts of Senecio jacobaea (syn. Jacobaea vulgaris, common or tansy ragwort) test positive in the Salmonella/microsome mutagenicity test (Ames test). Pyrrolizidine alkaloids (PAs) are thought to be responsible for these mutagenic
Kristine Opsvik Wikene et al.
Drug development and industrial pharmacy, 41(6), 969-977 (2014-05-21)
Bacterial resistance to antibiotics is increasing and alternative antibacterial treatments like antimicrobial photodynamic therapy (aPDT) are needed. Curcumin is under investigation as a potential photosensitizer in aPDT. The purpose of this study was to develop rapidly dissolving formulations of curcumin
Natasha R Ryz et al.
American journal of physiology. Gastrointestinal and liver physiology, 309(9), G730-G742 (2015-09-05)
Vitamin D deficiency affects more that 1 billion people worldwide. Although thought to increase risk of bacterial infections, the importance of vitamin D on host defense against intestinal bacterial pathogens is currently unclear since injection of the active form of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service