8.01600
4-Bromoaniline
for synthesis
Synonym(s):
4-Bromoaniline, p-Bromoaniline, p-Bromophenylamine
About This Item
Recommended Products
Quality Level
Assay
≥98.0% (GC)
form
solid
potency
456 mg/kg LD50, oral (Rat)
536 mg/kg LD50, skin (Rat)
bp
230-250 °C/1013 hPa
mp
60-64 °C
bulk density
810 kg/m3
storage temp.
2-30°C
SMILES string
Brc1ccc(cc1)N
InChI
1S/C6H6BrN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2
InChI key
WDFQBORIUYODSI-UHFFFAOYSA-N
Related Categories
Analysis Note
Melting range (lower value): ≥ 60 °C
Melting range (upper value): ≤ 64 °C
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service