Skip to Content
Merck
All Photos(1)

Key Documents

178496

Sigma-Aldrich

Apoptosis Activator VI, CD437/AHPN

The Apoptosis Activator VI, CD437/AHPN, also referenced under CAS 125316-60-1, modulates Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Apoptosis Activator VI, CD437/AHPN, 6-(3-(1-Adamantyl)-4-hydroxyphenyl)-2-naphthalene carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H26O3
CAS Number:
Molecular Weight:
398.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 39 mg/mL
ethanol: 39 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Oc1c(cc(cc1)c5cc6c(cc(cc6)C(=O)O)cc5)C32CC4CC(C3)CC(C2)C4

InChI

1S/C27H26O3/c28-25-6-5-22(20-1-2-21-11-23(26(29)30)4-3-19(21)10-20)12-24(25)27-13-16-7-17(14-27)9-18(8-16)15-27/h1-6,10-12,16-18,28H,7-9,13-15H2,(H,29,30)

InChI key

LDGIHZJOIQSHPB-UHFFFAOYSA-N

General description

A cell-permeable retinoid-related molecule that induces tumor growth arrest and apoptosis. The exact molecular basis of the underlying mechanism(s) is not yet well understood, however, studies using ovarian adenocarcinoma cells have shown that CD437/AHPN effectively functions as an ER stress inducer.

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 months at -20°C.

Other Notes

Watanabe, Y., et al. 2008. Biochem. Biophys. Res. Comm.366, 840.
Holmes, W.F., et al. 2004. J. Cell. Physiol.199, 317.
Zhao, X., and Spanjaard, R.A. 2003. J. Biomed. Sci.10, 44.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob Paiano et al.
Genes & development, 35(19-20), 1356-1367 (2021-09-11)
Double-strand break (DSB) repair choice is greatly influenced by the initial processing of DNA ends. 53BP1 limits the formation of recombinogenic single-strand DNA (ssDNA) in BRCA1-deficient cells, leading to defects in homologous recombination (HR). However, the exact mechanisms by which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service