Z423416
Easy-Read® thermometer
temperature -20-110 °C, immersion level, 76 mm
Synonym(s):
immersion thermometer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
feature
immersion level 76 mm
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60301-0200
temperature measuring range
-20-110 °C
L
305 mm
O.D.
6-7 mm
Looking for similar products? Visit Product Comparison Guide
General description
Easy-to-read thermometers with black printing on a bright yellow background. Filled with contrasting black, non-hazardous, Enviro-Safe liquid. Calibrated with NIST, DKD standards.
Application
Easy-Read® thermometer can be used in pharmaceutical, petrochemical, food, beverage processing, dairies, breweries, distilleries, universities and chemical labs.
Legal Information
Easy Read is a registered trademark of SP Scienceware, a division of SP
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service