Key Documents
Z261564
Luer-to-Luer connector
Micro-Mate® female Luer to male Luer, nickel plated
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
material
nickel plated
manufacturer/tradename
Popper 6414
Related Categories
General description
Water tested 45 psi.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)