DWKKC92812G-100
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with KIMCOTE® safety coating and glass stopper
borosilicate glass flask, flask capacity (100 mL), class A
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass flask
feature
class A
packaging
case of 6 ea
manufacturer/tradename
Kimble® KC92812G-100
flask capacity
(100 mL)
Related Categories
General description
KimCote Class A Heavy Duty Wide Mouth Volumetric Flasks
- Pipet access is easy with wide-mouth volumetric flasks
- Superior strength, durability and safety from heavy, uniform walls
- Quick identification with large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Standard Taper glass stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
KIMCOTE is a registered trademark of DWK LIFE SCIENCES LLC
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)