Key Documents
CLS49605L
Pyrex® round-bottom three neck vertical unequal height flask
capacity 5,000 mL, center joint: ST/NS 45/40
Synonym(s):
bottom flask, flask round, flask round bottom
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
three-neck (Vertical)
Joint
center joint: ST/NS45/40
side joint ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
case of 3
manufacturer/tradename
Corning 4960-5L
capacity
5,000 mL
General description
With full length joints.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)