Documents
BR701928
BRAND® replacement piston rod for TRANSFERPETTOR
for fixed type 1, 2, 3, 4, 5 μL
Synonym(s):
Accessories, Pipettes, Piston rod, Piston-operated pipettes, Positive displacement principle, Transferpettor
About This Item
UNSPSC Code:
41121501
NACRES:
NB.01
material
stainless steel
packaging
pack of 3 ea
manufacturer/tradename
BRAND 701928
General description
The BRAND® Replacement Piston Rod for the TRANSFERPETTOR is designed to ensure optimal performance and precision in liquid handling. Starting from 20 µl, product comes equipped with a seal. The replacement rod is easy to install and maintain, providing a reliable solution for users seeking to extend the life of their TRANSFERPETTOR while maintaining accuracy and efficiency in their pipetting tasks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG