BAH186101440
Stak-A-Tray™ large tray
W × L 35.6 cm × 35.6 cm
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H18610-1440
W × L
35.6 cm × 35.6 cm
General description
Modular rack and tray system allows multi-layer storage and prevents tipping of stacked flasks that result in spills.
Features and Benefits
- Can be used with Petri dishes and cell culture flasks in incubators, cold rooms or on the bench
- Each tier accommodates one large or two small trays
- Stack up to five racks for increased storage within the same footprint
- Perforated trays allow air flow and slide easily in the rack
- All parts are stainless steel; autoclavable at 121ºC (250ºF)
- Racks and trays sold separately
Legal Information
Stak-A-Tray is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)