Skip to Content
Merck
All Photos(1)

Documents

280135

Sigma-Aldrich

2-[4-(Dimethylamino)styryl]-1-methylpyridinium iodide

Dye content 95 %

Synonym(s):

2-Di-1-ASP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19IN2
CAS Number:
Molecular Weight:
366.24
Beilstein:
3776369
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 95%

mp

280 °C (dec.) (lit.)

λmax

466 nm

fluorescence

λex 466 nm; λem 504 nm in methanol
λex 475 nm; λem 589 nm in methanol

SMILES string

[I-].CN(C)c1ccc(cc1)\C=C\c2cccc[n+]2C

InChI

1S/C16H19N2.HI/c1-17(2)15-10-7-14(8-11-15)9-12-16-6-4-5-13-18(16)3;/h4-13H,1-3H3;1H/q+1;/p-1

InChI key

XPOIQAIBZGSIDD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

2-[4-(Dimethylamino)styryl]-1-methylpyridinium iodide (DASPMI) is a mitochondria staining styryl dye that is used to generate a quick responsive mechanism by applying changes in transmembrane potential. DASPMI can be used as a low toxic stain that shows a fluorescent behavior for mitochondria.
2-[4-(Dimethylamino)styryl]-1-methylpyridinium iodide (DASPMI) is a styrylcyanine dye.

Application

DASPMI is a polar sensitive dye that measures the membrane potential of mitochondria in living cells. It may be used as a light absorber in the development of a photoinitiator based system.
DASPMI is popularly used in selective staining of mitochondria in living cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distribution pattern of viable mitochondria in bovine lens epithelial cells.
O Leip et al.
Developments in ophthalmology, 26, 90-96 (1994-01-01)
Use of fluorescent probes to investigate the metabolic state of Pneumocystis carinii mitochondria.
F Chen et al.
The Journal of eukaryotic microbiology, 41(5), 79S-79S (1994-09-01)
Microtubules mediate mitochondrial distribution in fission yeast
Yaffe MP, et al.
Proceedings of the National Academy of Sciences of the USA, 93, 11664-11668 (1996)
Dibakar Sahoo et al.
The journal of physical chemistry. B, 114(5), 2044-2050 (2010-01-22)
The mode of binding of 2-(4-(dimethylamino)styryl)-1-methylpyridinium iodide (DASPMI) with calf thymus DNA as revealed from different steady state and time-resolved emission spectroscopic measurements has been reported in this paper. Fluorescence enhancement of DASPMI and its quenching by potassium iodide (KI)
J W Li et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 29(4), 316-319 (1994-01-01)
A reversed phase high performance liquid chromatography (RP-HPLC) method for determination of 2-(p-dimethylaminostyryl) pyridine methiodide (DSPM) concentration in rabbit plasma is reported. The stationary phase was YWG-C18H37. The mobile phase consisted of 80% CH3OH, 20% CH3COOH-CH3COONa buffer (pH 4.0, 0.20

Articles

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service